missing translation for 'onlineSavingsMsg'
Learn More
Learn More
2-Methyl-1,5-diaminopentane 98.0+%, TCI America™
$50.33 - $96.94
Chemical Identifiers
| CAS | 15520-10-2 |
|---|---|
| Molecular Formula | C6H16N2 |
| Molecular Weight (g/mol) | 116.208 |
| MDL Number | MFCD00013460 |
| InChI Key | JZUHIOJYCPIVLQ-UHFFFAOYSA-N |
| Synonym | 1,5-Diamino-2-methylpentane, 2-Methylpentamethylenediamine, 2-Methyl-1,5-pentanediamine |
| PubChem CID | 85862 |
| IUPAC Name | 2-methylpentane-1,5-diamine |
| SMILES | CC(CCCN)CN |
| Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
|---|---|---|---|---|---|---|---|---|---|
| Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
|
M020525ML
|
TCI America
M020525ML |
25 mL |
Each for $50.33
|
|
|||||
|
M0205500ML
|
TCI America
M0205500ML |
500 mL |
Each for $96.94
|
|
|||||
Chemical Identifiers
| 15520-10-2 | |
| 116.208 | |
| JZUHIOJYCPIVLQ-UHFFFAOYSA-N | |
| 85862 | |
| CC(CCCN)CN |
| C6H16N2 | |
| MFCD00013460 | |
| 1,5-Diamino-2-methylpentane, 2-Methylpentamethylenediamine, 2-Methyl-1,5-pentanediamine | |
| 2-methylpentane-1,5-diamine |
Specifications
| 15520-10-2 | |
| 78°C | |
| MFCD00013460 | |
| 2735 | |
| JZUHIOJYCPIVLQ-UHFFFAOYSA-N | |
| 2-methylpentane-1,5-diamine | |
| 85862 | |
| ≥98.0% (GC,T) | |
| 2-Methyl-1,5-diaminopentane |
| Yellow | |
| C6H16N2 | |
| 25 mL | |
| 1,5-Diamino-2-methylpentane, 2-Methylpentamethylenediamine, 2-Methyl-1,5-pentanediamine | |
| CC(CCCN)CN | |
| 116.208 | |
| 116.21 | |
| Liquid |
Safety and Handling
EINECSNumber : (2)-3724
RTECSNumber : SA0248500
TSCA : Yes