missing translation for 'onlineSavingsMsg'
Learn More

2-Methyl-1,5-diaminopentane 98.0+%, TCI America™

Supplier:  TCI America M020525ML

 View more versions of this product

Catalog No. M020525ML


Only null left
Explore available promotions

Description

Description

Specifications

Specifications

2-Methyl-1,5-diaminopentane
Yellow
C6H16N2
25 mL
1,5-Diamino-2-methylpentane, 2-Methylpentamethylenediamine, 2-Methyl-1,5-pentanediamine
CC(CCCN)CN
116.208
116.21
Liquid
15520-10-2
78°C
MFCD00013460
2735
JZUHIOJYCPIVLQ-UHFFFAOYSA-N
2-methylpentane-1,5-diamine
85862
≥98.0% (GC,T)

Chemical Identifiers

15520-10-2
116.208
JZUHIOJYCPIVLQ-UHFFFAOYSA-N
85862
CC(CCCN)CN
C6H16N2
MFCD00013460
1,5-Diamino-2-methylpentane, 2-Methylpentamethylenediamine, 2-Methyl-1,5-pentanediamine
2-methylpentane-1,5-diamine
Product Suggestions

Product Suggestions

Videos
Safety and Handling

Safety and Handling

EINECSNumber : (2)-3724

RTECSNumber : SA0248500

TSCA : Yes

SDS
Documents

Documents

Promotions

Promotions