missing translation for 'onlineSavingsMsg'
Learn More
Learn More
2-Methyl-1,5-diaminopentane 98.0+%, TCI America™
Supplier: TCI America M0205500ML
Specifications
| 2-Methyl-1,5-diaminopentane | |
| Yellow | |
| C6H16N2 | |
| 500 mL | |
| 1,5-Diamino-2-methylpentane, 2-Methylpentamethylenediamine, 2-Methyl-1,5-pentanediamine | |
| CC(CCCN)CN | |
| 116.208 | |
| 116.21 | |
| Liquid |
| 15520-10-2 | |
| 78°C | |
| MFCD00013460 | |
| 2735 | |
| JZUHIOJYCPIVLQ-UHFFFAOYSA-N | |
| 2-methylpentane-1,5-diamine | |
| 85862 | |
| ≥98.0% (GC,T) |
Chemical Identifiers
| 15520-10-2 | |
| 116.208 | |
| JZUHIOJYCPIVLQ-UHFFFAOYSA-N | |
| 85862 | |
| CC(CCCN)CN |
| C6H16N2 | |
| MFCD00013460 | |
| 1,5-Diamino-2-methylpentane, 2-Methylpentamethylenediamine, 2-Methyl-1,5-pentanediamine | |
| 2-methylpentane-1,5-diamine |
Safety and Handling
EINECSNumber : (2)-3724
RTECSNumber : SA0248500
TSCA : Yes