missing translation for 'onlineSavingsMsg'
Learn More
Learn More
2-Methyl-1,4-naphthoquinone 98.0+%, TCI America™
Supplier: TCI America M0373250G
Specifications
| 2-Methyl-1,4-naphthoquinone | |
| 107°C | |
| MFCD00001681 | |
| menadione, vitamin k3, 2-methyl-1,4-naphthoquinone, menaphthone, 2-methylnaphthoquinone, thyloquinone, kappaxin, kayquinone, klottone, panosine | |
| CC1=CC(=O)C2=CC=CC=C2C1=O | |
| 172.183 | |
| CHEBI:28869 | |
| ≥98.0% (HPLC,T) |
| 58-27-5 | |
| C11H8O2 | |
| 250 g | |
| MJVAVZPDRWSRRC-UHFFFAOYSA-N | |
| 2-methylnaphthalene-1,4-dione | |
| 4055 | |
| 172.18 | |
| Crystalline Powder |
Chemical Identifiers
| 58-27-5 | |
| 172.183 | |
| MJVAVZPDRWSRRC-UHFFFAOYSA-N | |
| 4055 | |
| 2-methylnaphthalene-1,4-dione |
| C11H8O2 | |
| MFCD00001681 | |
| menadione, vitamin k3, 2-methyl-1,4-naphthoquinone, menaphthone, 2-methylnaphthoquinone, thyloquinone, kappaxin, kayquinone, klottone, panosine | |
| CHEBI:28869 | |
| CC1=CC(=O)C2=CC=CC=C2C1=O |
Safety and Handling
EINECSNumber : (4)-0373
RTECSNumber : QL9100000
TSCA : Yes