missing translation for 'onlineSavingsMsg'
Learn More

2-Methyl-1,4-naphthoquinone 98.0+%, TCI America™

Catalog No. p-7132128
Change view
missing translation for 'orderingAttributeHoverText'
Quantity:
25 g
250 g
Catalog No. Quantity
M037325G 25 g
M0373250G 250 g
2 options

Catalog No. M037325G

Supplier: TCI America M037325G

Only null left

Chemical Identifiers

CAS 58-27-5
Molecular Formula C11H8O2
Molecular Weight (g/mol) 172.183
MDL Number MFCD00001681
InChI Key MJVAVZPDRWSRRC-UHFFFAOYSA-N
Synonym menadione, vitamin k3, 2-methyl-1,4-naphthoquinone, menaphthone, 2-methylnaphthoquinone, thyloquinone, kappaxin, kayquinone, klottone, panosine
PubChem CID 4055
ChEBI CHEBI:28869
IUPAC Name 2-methylnaphthalene-1,4-dione
SMILES CC1=CC(=O)C2=CC=CC=C2C1=O

Specifications

Melting Point 107°C
Quantity 25 g
Formula Weight 172.18
Percent Purity ≥98.0% (HPLC,T)
Physical Form Crystalline Powder
Chemical Name or Material 2-Methyl-1,4-naphthoquinone
EINECSNumber (4)-0373
RTECSNumber QL9100000
TSCA Yes