missing translation for 'onlineSavingsMsg'
Learn More

2-Methoxytoluene 98.0+%, TCI America™

Catalog No. M015025ML
Change view
Click to view available options
Quantity:
25 mL
500 mL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
M015025ML 25 mL
M0150500ML 500 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. M015025ML Supplier TCI America Supplier No. M015025ML

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 578-58-5
Molecular Formula C8H10O
Molecular Weight (g/mol) 122.167
MDL Number MFCD00008373
InChI Key DTFKRVXLBCAIOZ-UHFFFAOYSA-N
Synonym 2-methylanisole, o-methylanisole, 2-methoxytoluene, o-cresol methyl ether, o-cresyl methyl ether, anisole, o-methyl, benzene, 1-methoxy-2-methyl, methyl o-tolyl ether, o-methoxytoluene, 2-methylmethoxybenzene
PubChem CID 33637
IUPAC Name 1-methoxy-2-methylbenzene
SMILES CC1=CC=CC=C1OC

Specifications

Color Yellow
Boiling Point 171°C
Quantity 25 mL
UN Number 3271
Formula Weight 122.17
Percent Purity ≥98.0% (GC)
Physical Form Liquid
Chemical Name or Material 2-Methoxytoluene
EINECSNumber (3)-0573
TSCA Yes