missing translation for 'onlineSavingsMsg'
Learn More

2-Methoxythiophenol, 97%

Catalog No. AAL0968906
missing translation for 'changeView'
missing translation for 'orderingAttributeHoverText'
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAL0968906 5 g
AAL0968914 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 missing translation for 'options'
Catalog No. AAL0968906 missing translation for 'mfr' Thermo Scientific Chemicals missing translation for 'supplierNo' L0968906

CAS: 7217-59-6 | C7H8OS | 140.2 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 7217-59-6
Molecular Formula C7H8OS
Molecular Weight (g/mol) 140.2
MDL Number MFCD00004834
InChI Key DSCJETUEDFKYGN-UHFFFAOYSA-N
Synonym 2-methoxythiophenol, thioguaiacol, benzenethiol, 2-methoxy, o-methoxythiophenol, o-methoxybenzenethiol, 2-methoxy thiophenol, 2-mercaptoanisole, 2-methoxybenzene-1-thiol, 2-methoxy-benzenethiol, benzenethiol, o-methoxy
PubChem CID 23642
IUPAC Name 2-methoxybenzenethiol
SMILES COC1=CC=CC=C1S

Specifications

Density 1.152
Boiling Point 225°C to 228°C
Flash Point 85°C (185°F)
Odor Unpleasant
Refractive Index 1.5918
Quantity 5 g
Beilstein 2042178
Sensitivity Air sensitive
Formula Weight 140.21
Percent Purity 97%
Chemical Name or Material 2-Methoxythiophenol
Show More Show Less
missing translation for 'healthHazard1' H302-H315-H319-H335
missing translation for 'healthHazard2' GHS H Statement
H302-H315-H319-H335-H227
Harmful if swallowed.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Combustible liquid.
missing translation for 'healthHazard3' P261-P264b-P270-P271-P280-P301+P312-P302+P352-P304+P340-P305+P351+P338-P312-P330-P332+P313-P362-P501c
EINECSNumber 230-605-7
RTECSNumber DC1790000
TSCA Yes
Recommended Storage Ambient temperatures; Store under Argon

RUO – Research Use Only