accessibility menu, dialog, popup

UserName

H3COHS

CAS RN 7217-59-6

IUPAC Name: 2-methoxybenzene-1-thiol
Synonyms: thioguaiacol
Molecular Weight (g/mol): 140.2
Molecular Formula: C7H8OS
InChi Key: DSCJETUEDFKYGN-UHFFFAOYSA-N
SMILES: COC1=CC=CC=C1S
Boiling Point 
  • (2)
  • (2)
Percent Purity 
  • (2)
  • (2)
Quantity 
  • (2)
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (2)
  • (2)
  • (2)
  • (2)
  • (2)
  • (2)
  • (2)
  • (2)
  • (2)