missing translation for 'onlineSavingsMsg'
Learn More

2-Methoxythiobenzamide, 97%

Catalog No. p-7031605
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAH3273703 1 g
AAH3273706 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAH3273703 Supplier Thermo Scientific Chemicals Supplier No. H3273703
Only null left

CAS: 42590-97-6 | C8H9NOS | 167.226 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 42590-97-6
Molecular Formula C8H9NOS
Molecular Weight (g/mol) 167.226
MDL Number MFCD04973324
InChI Key YNEHBLLUZJTDOL-UHFFFAOYSA-N
PubChem CID 2759731
IUPAC Name 2-methoxybenzenecarbothioamide
SMILES COC1=CC=CC=C1C(=S)N

Specifications

Melting Point 109°C to 113°C
Quantity 1 g
Synonym 2-methoxythiobenzamide, 2-methoxybenzothioamide, 2-methoxy-thiobenzamide, 2-methoxybenzene-1-carbothioamide, amino 2-methoxyphenyl methane-1-thione, o-methoxythiobenzamide, 2-methoxy thiobenzamide, #, benzenecarbothioamide,2-methoxy, benzenecarbothioamide, 2-methoxy
Formula Weight 167.23
Percent Purity 97%
Chemical Name or Material 2-Methoxythiobenzamide
Hazard Category H302-H315-H319-H335
Hazard Statement GHS H Statement
H301-H315-H319-H335
Toxic if swallowed.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement P261-P264b-P270-P271-P280-P301+P312-P302+P352-P304+P340-P305+P351+P338-P312-P330-P332+P313-P362-P501c
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only