accessibility menu, dialog, popup

UserName

CAS RN 42590-97-6

IUPAC Name: 2-methoxybenzene-1-carbothioamide
Synonyms: 2-methoxybenzenecarbothioamide
Molecular Weight (g/mol): 167.23
Molecular Formula: C8H9NOS
InChi Key: YNEHBLLUZJTDOL-UHFFFAOYSA-N
SMILES: COC1=CC=CC=C1C(N)=S

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results