Learn More
2-Methoxypyridine-5-boronic acid, 95%
CAS: 163105-89-3 | C6H8BNO3 | 152.94 g/mol
Supplier: Thermo Scientific Chemicals L2008706
Description
2-Methoxypyridine-5-boronic acid is used in cosmetics, coatings candles.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications2-Methoxypyridine-5-boronic acid is used in cosmetics, coatings candles.
Solubility
Soluble in water.
Notes
Store away from strong oxidizing agents. Keep container tightly closed. Store in cool, dry conditions in well sealed containers
Specifications
| 2-Methoxypyridine-5-boronic acid | |
| 132°C to 133°C | |
| MFCD02093044 | |
| 9119950 | |
| Soluble in water. | |
| COC1=CC=C(C=N1)B(O)O | |
| 152.94 | |
| 152.95 |
| 163105-89-3 | |
| C6H8BNO3 | |
| 5 g | |
| 2-methoxy-5-pyridineboronic acid, 6-methoxypyridin-3-yl boronic acid, 2-methoxypyridine-5-boronic acid, 2-methoxy-5-pyridinylboronic acid, 6-methoxypyridine-3-boronic acid, 6-methoxypyridin-3-yl-3-boronic acid, 6-methoxy-pyridine-3-boronic acid, 6-methoxy-3-pyridinylboronic acid, 6-methoxy-3-pyridylboronic acid, 6-methoxypyridin-3-yl boranediol | |
| DHADXDMPEUWEAS-UHFFFAOYSA-N | |
| (6-methoxypyridin-3-yl)boronic acid | |
| 2734368 | |
| 95% |
Chemical Identifiers
| 163105-89-3 | |
| 152.94 | |
| DHADXDMPEUWEAS-UHFFFAOYSA-N | |
| 2734368 | |
| COC1=CC=C(C=N1)B(O)O |
| C6H8BNO3 | |
| MFCD02093044 | |
| 2-methoxy-5-pyridineboronic acid, 6-methoxypyridin-3-yl boronic acid, 2-methoxypyridine-5-boronic acid, 2-methoxy-5-pyridinylboronic acid, 6-methoxypyridine-3-boronic acid, 6-methoxypyridin-3-yl-3-boronic acid, 6-methoxy-pyridine-3-boronic acid, 6-methoxy-3-pyridinylboronic acid, 6-methoxy-3-pyridylboronic acid, 6-methoxypyridin-3-yl boranediol | |
| (6-methoxypyridin-3-yl)boronic acid |
Safety and Handling
GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
P261-P264b-P270-P271-P280-P301+P312-P302+P352-P304+P340-P305+P351+P338-P312-P330-P332+P313-P362-P501c
H302-H315-H319-H335
TSCA : No
Recommended Storage : Ambient temperatures
RUO – Research Use Only