missing translation for 'onlineSavingsMsg'
Learn More

2-Methoxypyridine-5-boronic acid, 95%

Catalog No. p-7048012
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAL2008703 1 g
AAL2008706 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAL2008703 Supplier Thermo Scientific Chemicals Supplier No. L2008703

CAS: 163105-89-3 | C6H8BNO3 | 152.94 g/mol

2-Methoxypyridine-5-boronic acid is used in cosmetics, coatings candles.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
2-Methoxypyridine-5-boronic acid is used in cosmetics, coatings candles.

Solubility
Soluble in water.

Notes
Store away from strong oxidizing agents. Keep container tightly closed. Store in cool, dry conditions in well sealed containers

Chemical Identifiers

CAS 163105-89-3
Molecular Formula C6H8BNO3
Molecular Weight (g/mol) 152.94
MDL Number MFCD02093044
InChI Key DHADXDMPEUWEAS-UHFFFAOYSA-N
Synonym 2-methoxy-5-pyridineboronic acid, 6-methoxypyridin-3-yl boronic acid, 2-methoxypyridine-5-boronic acid, 2-methoxy-5-pyridinylboronic acid, 6-methoxypyridine-3-boronic acid, 6-methoxypyridin-3-yl-3-boronic acid, 6-methoxy-pyridine-3-boronic acid, 6-methoxy-3-pyridinylboronic acid, 6-methoxy-3-pyridylboronic acid, 6-methoxypyridin-3-yl boranediol
PubChem CID 2734368
IUPAC Name (6-methoxypyridin-3-yl)boronic acid
SMILES COC1=CC=C(C=N1)B(O)O

Specifications

Melting Point 132°C to 133°C
Quantity 1 g
Beilstein 9119950
Solubility Information Soluble in water.
Formula Weight 152.95
Percent Purity 95%
Chemical Name or Material 2-Methoxypyridine-5-boronic acid
Hazard Category H302-H315-H319-H335
Hazard Statement GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement P261-P264b-P270-P271-P280-P301+P312-P302+P352-P304+P340-P305+P351+P338-P312-P330-P332+P313-P362-P501c
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only