accessibility menu, dialog, popup

UserName

H3CONBOHHO

CAS RN 163105-89-3

IUPAC Name: (6-methoxypyridin-3-yl)boronic acid
Synonyms: 6-methoxypyridin-3-ylboronic acid
Molecular Weight (g/mol): 152.94
Molecular Formula: C6H8BNO3
InChi Key: DHADXDMPEUWEAS-UHFFFAOYSA-N
SMILES: COC1=CC=C(C=N1)B(O)O
Quantity 
  • (2)
  • (1)
Physical Form 
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (1)
  • (2)
  • (2)
  • (1)
  • (1)