missing translation for 'onlineSavingsMsg'
Learn More

2-Methoxyphenyl Isothiocyanate 98.0+%, TCI America™

Catalog No. I05125G
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
I05125G 5 g
I051225G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. I05125G Supplier TCI America Supplier No. I05125G

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 3288-04-8
Molecular Formula C8H7NOS
Molecular Weight (g/mol) 165.21
MDL Number MFCD00011675
InChI Key QKAOOWJWWKWWOZ-UHFFFAOYSA-N
Synonym 2-methoxyphenyl isothiocyanate, benzene, 1-isothiocyanato-2-methoxy, o-methoxyphenyl isothiocyanate, 2-methoxyphenylisothiocyanate, 1-isothiocyanato-2-methoxy-benzene, isothiocyanic acid 2-methoxyphenyl ester, 2-methoxybenzenisothiocyanate, acmc-1coqu, o-methoxyphenylisothiocyanat, orthomethoxyphenyl isothiocyanate
PubChem CID 520599
IUPAC Name 1-isothiocyanato-2-methoxybenzene
SMILES COC1=CC=CC=C1N=C=S

Specifications

Color Yellow
Boiling Point 267°C
Quantity 5 g
UN Number 1760
Formula Weight 165.21
Percent Purity ≥98.0% (GC)
Physical Form Liquid
Chemical Name or Material 2-Methoxyphenyl Isothiocyanate