accessibility menu, dialog, popup

UserName

CH3ONCS

CAS RN 3288-04-8

IUPAC Name: 1-isothiocyanato-2-methoxybenzene
Synonyms: 1-isothiocyanato-2-methoxybenzene
Molecular Weight (g/mol): 165.21
Molecular Formula: C8H7NOS
InChi Key: QKAOOWJWWKWWOZ-UHFFFAOYSA-N
SMILES: COC1=CC=CC=C1N=C=S
Molecular Weight (g/mol) 
  • (1)
  • (3)
Percent Purity 
  • (2)
  • (2)
Quantity 
  • (2)
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (2)
  • (2)
  • (4)
  • (1)
  • (3)
  • (2)
  • (2)
  • (2)
  • (2)
  • (2)