missing translation for 'onlineSavingsMsg'
Learn More
Learn More
2-Methoxyphenyl Isocyanate 98.0+%, TCI America™
$61.51 - $188.27
Chemical Identifiers
| CAS | 700-87-8 |
|---|---|
| Molecular Formula | C8H7NO2 |
| Molecular Weight (g/mol) | 149.15 |
| MDL Number | MFCD00002004 |
| InChI Key | SUVCZZADQDCIEQ-UHFFFAOYSA-N |
| Synonym | 2-methoxyphenyl isocyanate, 2-methoxyphenylisocyanate, benzene, 1-isocyanato-2-methoxy, o-methoxyphenyl isocyanate, 2-isocyanatoanisole, 1-isocyanato-2-methoxy-benzene, o-anisylisocyanate, 2-methoxybenzenisocyanate, o-anisyl isocyanate, methoxyphenyl isocyanate |
| PubChem CID | 69695 |
| IUPAC Name | 1-isocyanato-2-methoxybenzene |
| SMILES | COC1=CC=CC=C1N=C=O |
Chemical Identifiers
| 700-87-8 | |
| 149.15 | |
| SUVCZZADQDCIEQ-UHFFFAOYSA-N | |
| 69695 | |
| COC1=CC=CC=C1N=C=O |
| C8H7NO2 | |
| MFCD00002004 | |
| 2-methoxyphenyl isocyanate, 2-methoxyphenylisocyanate, benzene, 1-isocyanato-2-methoxy, o-methoxyphenyl isocyanate, 2-isocyanatoanisole, 1-isocyanato-2-methoxy-benzene, o-anisylisocyanate, 2-methoxybenzenisocyanate, o-anisyl isocyanate, methoxyphenyl isocyanate | |
| 1-isocyanato-2-methoxybenzene |
Specifications
| 700-87-8 | |
| 96°C | |
| MFCD00002004 | |
| 2206 | |
| SUVCZZADQDCIEQ-UHFFFAOYSA-N | |
| 1-isocyanato-2-methoxybenzene | |
| 69695 | |
| ≥98.0% (GC) | |
| 2-Methoxyphenyl Isocyanate |
| Yellow | |
| C8H7NO2 | |
| 5 g | |
| 2-methoxyphenyl isocyanate, 2-methoxyphenylisocyanate, benzene, 1-isocyanato-2-methoxy, o-methoxyphenyl isocyanate, 2-isocyanatoanisole, 1-isocyanato-2-methoxy-benzene, o-anisylisocyanate, 2-methoxybenzenisocyanate, o-anisyl isocyanate, methoxyphenyl isocyanate | |
| COC1=CC=CC=C1N=C=O | |
| 149.15 | |
| 149.15 | |
| Liquid |
Safety and Handling
TSCA : Yes
Recommended Storage : Refrigerator