accessibility menu, dialog, popup

UserName

CH3ONCO

CAS RN 700-87-8

IUPAC Name: 1-isocyanato-2-methoxybenzene
Synonyms: 1-isocyanato-2-methoxybenzene
Molecular Weight (g/mol): 149.15
Molecular Formula: C8H7NO2
InChi Key: SUVCZZADQDCIEQ-UHFFFAOYSA-N
SMILES: COC1=CC=CC=C1N=C=O
Boiling Point 
  • (2)
  • (2)
Percent Purity 
  • (2)
  • (2)
Quantity 
  • (2)
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (2)
  • (2)
  • (2)
  • (2)
  • (2)
  • (2)
  • (2)
  • (2)
  • (2)