missing translation for 'onlineSavingsMsg'
Learn More

2-Methoxyphenyl Isocyanate 98.0+%, TCI America™

Catalog No. I04395G
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
I04395G 5 g
I043925G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. I04395G Supplier TCI America Supplier No. I04395G

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 700-87-8
Molecular Formula C8H7NO2
Molecular Weight (g/mol) 149.15
MDL Number MFCD00002004
InChI Key SUVCZZADQDCIEQ-UHFFFAOYSA-N
Synonym 2-methoxyphenyl isocyanate, 2-methoxyphenylisocyanate, benzene, 1-isocyanato-2-methoxy, o-methoxyphenyl isocyanate, 2-isocyanatoanisole, 1-isocyanato-2-methoxy-benzene, o-anisylisocyanate, 2-methoxybenzenisocyanate, o-anisyl isocyanate, methoxyphenyl isocyanate
PubChem CID 69695
IUPAC Name 1-isocyanato-2-methoxybenzene
SMILES COC1=CC=CC=C1N=C=O

Specifications

Color Yellow
Boiling Point 96°C
Quantity 5 g
UN Number 2206
Formula Weight 149.15
Percent Purity ≥98.0% (GC)
Physical Form Liquid
Chemical Name or Material 2-Methoxyphenyl Isocyanate
TSCA Yes
Recommended Storage Refrigerator