Learn More
2-Methoxyphenyl isocyanate, 98%
CAS: 700-87-8 | C8H7NO2 | 149.15 g/mol
$75.19 - $236.55
Chemical Identifiers
| CAS | 700-87-8 |
|---|---|
| Molecular Formula | C8H7NO2 |
| Molecular Weight (g/mol) | 149.15 |
| MDL Number | MFCD00002004 |
| InChI Key | SUVCZZADQDCIEQ-UHFFFAOYSA-N |
| Synonym | 2-methoxyphenyl isocyanate, 2-methoxyphenylisocyanate, benzene, 1-isocyanato-2-methoxy, o-methoxyphenyl isocyanate, 2-isocyanatoanisole, 1-isocyanato-2-methoxy-benzene, o-anisylisocyanate, 2-methoxybenzenisocyanate, o-anisyl isocyanate, methoxyphenyl isocyanate |
| PubChem CID | 69695 |
| IUPAC Name | 1-isocyanato-2-methoxybenzene |
| SMILES | COC1=CC=CC=C1N=C=O |
| Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
|---|---|---|---|---|---|---|---|---|---|
| Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
|
AAA1807506
|
Thermo Scientific Chemicals
A1807506 |
5 g |
Each for $75.19
|
|
|||||
|
AAA1807514
|
Thermo Scientific Chemicals
A1807514 |
25 g |
Each for $236.55
|
|
|||||
Description
2-Methoxyphenyl isocyanate is an important raw material and intermediate used in organic synthesis, pharmaceuticals, agrochemicals and dyestuff.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications2-Methoxyphenyl isocyanate is an important raw material and intermediate used in organic synthesis, pharmaceuticals, agrochemicals and dyestuff.
Solubility
Hydrolyzes with water.
Notes
Moisture Sensitive. Protect from water and humidity. Store away from oxidizing agents. Store in a cool, dry conditions in a well sealed container. Store under dry inert gas.
Chemical Identifiers
| 700-87-8 | |
| 149.15 | |
| SUVCZZADQDCIEQ-UHFFFAOYSA-N | |
| 69695 | |
| COC1=CC=CC=C1N=C=O |
| C8H7NO2 | |
| MFCD00002004 | |
| 2-methoxyphenyl isocyanate, 2-methoxyphenylisocyanate, benzene, 1-isocyanato-2-methoxy, o-methoxyphenyl isocyanate, 2-isocyanatoanisole, 1-isocyanato-2-methoxy-benzene, o-anisylisocyanate, 2-methoxybenzenisocyanate, o-anisyl isocyanate, methoxyphenyl isocyanate | |
| 1-isocyanato-2-methoxybenzene |
Specifications
| 700-87-8 | |
| 94°C to 96°C (10 mmHg) | |
| Pungent | |
| 1.5414 | |
| 5 g | |
| 387700 | |
| 2-methoxyphenyl isocyanate, 2-methoxyphenylisocyanate, benzene, 1-isocyanato-2-methoxy, o-methoxyphenyl isocyanate, 2-isocyanatoanisole, 1-isocyanato-2-methoxy-benzene, o-anisylisocyanate, 2-methoxybenzenisocyanate, o-anisyl isocyanate, methoxyphenyl isocyanate | |
| SUVCZZADQDCIEQ-UHFFFAOYSA-N | |
| 1-isocyanato-2-methoxybenzene | |
| 69695 | |
| 98% |
| 1.122 | |
| >110°C (230°F) | |
| C8H7NO2 | |
| MFCD00002004 | |
| UN2206 | |
| Moisture sensitive | |
| Hydrolyzes with water. | |
| COC1=CC=CC=C1N=C=O | |
| 149.15 | |
| 149.15 | |
| 2-Methoxyphenyl isocyanate |
Safety and Handling
GHS H Statement
H331-H334-H302-H315-H319-H335
Toxic if inhaled.
May cause allergy or asthma symptoms or breathing difficulties if inhaled.
Harmful if swallowed.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
P261-P264b-P270-P271-P280-P301+P312-P302+P352-P304+P340-P305+P351+P338-P312-P330-P332+P313-P362-P501c
H302+H312+H332-H315-H319-H335
DOTInformation : Transport Hazard Class: 6.1; Packing Group: III; Proper Shipping Name: ISOCYANATES, TOXIC, N.O.S.
EINECSNumber : 211-853-5
TSCA : Yes
Recommended Storage : Keep cold
RUO – Research Use Only