missing translation for 'onlineSavingsMsg'
Learn More

2-Methoxyphenyl isocyanate, 98%

Catalog No. AAA1807506
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAA1807506 5 g
AAA1807514 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAA1807506 Supplier Thermo Scientific Chemicals Supplier No. A1807506

CAS: 700-87-8 | C8H7NO2 | 149.15 g/mol

2-Methoxyphenyl isocyanate is an important raw material and intermediate used in organic synthesis, pharmaceuticals, agrochemicals and dyestuff.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
2-Methoxyphenyl isocyanate is an important raw material and intermediate used in organic synthesis, pharmaceuticals, agrochemicals and dyestuff.

Solubility
Hydrolyzes with water.

Notes
Moisture Sensitive. Protect from water and humidity. Store away from oxidizing agents. Store in a cool, dry conditions in a well sealed container. Store under dry inert gas.

Chemical Identifiers

CAS 700-87-8
Molecular Formula C8H7NO2
Molecular Weight (g/mol) 149.15
MDL Number MFCD00002004
InChI Key SUVCZZADQDCIEQ-UHFFFAOYSA-N
Synonym 2-methoxyphenyl isocyanate, 2-methoxyphenylisocyanate, benzene, 1-isocyanato-2-methoxy, o-methoxyphenyl isocyanate, 2-isocyanatoanisole, 1-isocyanato-2-methoxy-benzene, o-anisylisocyanate, 2-methoxybenzenisocyanate, o-anisyl isocyanate, methoxyphenyl isocyanate
PubChem CID 69695
IUPAC Name 1-isocyanato-2-methoxybenzene
SMILES COC1=CC=CC=C1N=C=O

Specifications

Density 1.122
Boiling Point 94°C to 96°C (10 mmHg)
Flash Point >110°C (230°F)
Odor Pungent
Refractive Index 1.5414
Quantity 5 g
UN Number UN2206
Beilstein 387700
Sensitivity Moisture sensitive
Solubility Information Hydrolyzes with water.
Formula Weight 149.15
Percent Purity 98%
Chemical Name or Material 2-Methoxyphenyl isocyanate
Show More Show Less
Hazard Category H302+H312+H332-H315-H319-H335
Hazard Statement GHS H Statement
H331-H334-H302-H315-H319-H335
Toxic if inhaled.
May cause allergy or asthma symptoms or breathing difficulties if inhaled.
Harmful if swallowed.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement P261-P264b-P270-P271-P280-P301+P312-P302+P352-P304+P340-P305+P351+P338-P312-P330-P332+P313-P362-P501c
DOTInformation Transport Hazard Class: 6.1; Packing Group: III; Proper Shipping Name: ISOCYANATES, TOXIC, N.O.S.
EINECSNumber 211-853-5
TSCA Yes
Recommended Storage Keep cold

RUO – Research Use Only