missing translation for 'onlineSavingsMsg'
Learn More

2-Methoxybenzyl alcohol, 99%

CAS: 612-16-8 | C8H10O2 | 138.166 g/mol

$134.00 - $294.81

Chemical Identifiers

CAS 612-16-8
Molecular Formula C8H10O2
Molecular Weight (g/mol) 138.166
MDL Number MFCD00004611
InChI Key WYLYBQSHRJMURN-UHFFFAOYSA-N
Synonym 2-methoxybenzyl alcohol, 2-methoxyphenyl methanol, o-methoxybenzyl alcohol, benzenemethanol, 2-methoxy, o-anisyl alcohol, 2-methoxybenzylalcohol, 2-methoxybenzenemethanol, unii-457nq8ngcg, 457nq8ngcg, 2-methoxyphenyl methan-1-ol
PubChem CID 69154
IUPAC Name (2-methoxyphenyl)methanol
SMILES COC1=CC=CC=C1CO
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
AAA1401614
View Documents
Thermo Scientific Chemicals
A1401614
25 g
Each for $134.00
Only null left
 
AAA1401622
View Documents
Thermo Scientific Chemicals
A1401622
100 g
Each for $294.81
Only null left
 
Description

Description

It is an important raw material and intermediate used in Organic Synthesis, Pharmaceuticals, Agrochemicals.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
It is an important raw material and intermediate used in Organic Synthesis, Pharmaceuticals, Agrochemicals.

Solubility
Not miscible or difficult to mix in water. Soluble in ethanol.

Notes
Store in a cool, dry, well-ventilated area. Store away from oxidizing agent, strong acids.
Specifications

Chemical Identifiers

612-16-8
138.166
WYLYBQSHRJMURN-UHFFFAOYSA-N
69154
COC1=CC=CC=C1CO
C8H10O2
MFCD00004611
2-methoxybenzyl alcohol, 2-methoxyphenyl methanol, o-methoxybenzyl alcohol, benzenemethanol, 2-methoxy, o-anisyl alcohol, 2-methoxybenzylalcohol, 2-methoxybenzenemethanol, unii-457nq8ngcg, 457nq8ngcg, 2-methoxyphenyl methan-1-ol
(2-methoxyphenyl)methanol

Specifications

612-16-8
248°C to 250°C
C8H10O2
MFCD00004611
2043673
Not miscible or difficult to mix in water. Soluble in ethanol.
COC1=CC=CC=C1CO
138.166
138.17
2-Methoxybenzyl alcohol
1.12
>110°C (230°F)
1.5475
25 g
2-methoxybenzyl alcohol, 2-methoxyphenyl methanol, o-methoxybenzyl alcohol, benzenemethanol, 2-methoxy, o-anisyl alcohol, 2-methoxybenzylalcohol, 2-methoxybenzenemethanol, unii-457nq8ngcg, 457nq8ngcg, 2-methoxyphenyl methan-1-ol
WYLYBQSHRJMURN-UHFFFAOYSA-N
(2-methoxyphenyl)methanol
69154
99%
Videos
Safety and Handling

Safety and Handling

P261-P264b-P270-P271-P280-P301+P312-P302+P352-P304+P340-P305+P351+P338-P312-P330-P332+P313-P362-P501c

H302-H315-H319-H335

EINECSNumber : 210-296-5

TSCA : Yes

Recommended Storage : Ambient temperatures

SDS
Documents

Documents

RUO – Research Use Only