missing translation for 'onlineSavingsMsg'
Learn More
Learn More
2-Methoxybenzyl alcohol, 99%
Description
It is an important raw material and intermediate used in Organic Synthesis, Pharmaceuticals, Agrochemicals.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications
It is an important raw material and intermediate used in Organic Synthesis, Pharmaceuticals, Agrochemicals.
Solubility
Not miscible or difficult to mix in water. Soluble in ethanol.
Notes
Store in a cool, dry, well-ventilated area. Store away from oxidizing agent, strong acids.
It is an important raw material and intermediate used in Organic Synthesis, Pharmaceuticals, Agrochemicals.
Solubility
Not miscible or difficult to mix in water. Soluble in ethanol.
Notes
Store in a cool, dry, well-ventilated area. Store away from oxidizing agent, strong acids.
Specifications
Specifications
| Density | 1.12 |
| Boiling Point | 248°C to 250°C |
| Flash Point | >110°C (230°F) |
| Refractive Index | 1.5475 |
| Quantity | 100 g |
| Beilstein | 2043673 |
| Solubility Information | Not miscible or difficult to mix in water. Soluble in ethanol. |
| Formula Weight | 138.17 |
| Percent Purity | 99% |
| Chemical Name or Material | 2-Methoxybenzyl alcohol |
Safety and Handling
| Hazard Category | H302-H315-H319-H335 |
| Precautionary Statement | P261-P264b-P270-P271-P280-P301+P312-P302+P352-P304+P340-P305+P351+P338-P312-P330-P332+P313-P362-P501c |
| EINECSNumber | 210-296-5 |
| TSCA | Yes |
| Recommended Storage | Ambient temperatures |
RUO – Research Use Only