missing translation for 'onlineSavingsMsg'
Learn More

2-Methoxybenzenethiol 98.0+%, TCI America™

Catalog No. p-7132799
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
M16705G 5 g
M167025G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. M16705G Supplier TCI America Supplier No. M16705G
Only null left

Chemical Identifiers

CAS 7217-59-6
Molecular Formula C7H8OS
Molecular Weight (g/mol) 140.2
InChI Key DSCJETUEDFKYGN-UHFFFAOYSA-N
Synonym 2-methoxythiophenol, thioguaiacol, benzenethiol, 2-methoxy, o-methoxythiophenol, o-methoxybenzenethiol, 2-methoxy thiophenol, 2-mercaptoanisole, 2-methoxybenzene-1-thiol, 2-methoxy-benzenethiol, benzenethiol, o-methoxy
PubChem CID 23642
IUPAC Name 2-methoxybenzenethiol
SMILES COC1=CC=CC=C1S

Specifications

Color Yellow
Boiling Point 219°C
Quantity 5 g
Formula Weight 140.20
Percent Purity ≥98.0% (GC)
Physical Form Liquid
Chemical Name or Material 2-Methoxybenzenethiol
RTECSNumber DC1790000
TSCA Yes