missing translation for 'onlineSavingsMsg'
Learn More

2-Methoxy-6-nitrotoluene 97.0+%, TCI America™

Catalog No. p-7132717
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
M14165G 5 g
M141625G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. M14165G Supplier TCI America Supplier No. M14165G

Chemical Identifiers

CAS 4837-88-1
Molecular Formula C8H9NO3
Molecular Weight (g/mol) 167.16
MDL Number MFCD00007159
InChI Key HQCZLEAGIOIIMC-UHFFFAOYSA-N
Synonym 2-methyl-3-nitroanisole, 2-methoxy-6-nitrotoluene, benzene, 1-methoxy-2-methyl-3-nitro, 1-methoxy-2-methyl-3-nitro-benzene, anisole, 2-methyl-3-nitro, 6-methoxy-2-nitrotoluene, 1-meo-2-me-3-no2-benzene, 3-nitro-2-methyl anisole, 3-methoxy-2-methyl-1-nitrobenzene, 2-methyl-3-nitroanisole 2-methoxy-6-nitrotoluene
PubChem CID 78554
IUPAC Name 1-methoxy-2-methyl-3-nitrobenzene
SMILES COC1=CC=CC(=C1C)[N+]([O-])=O

Specifications

Melting Point 56°C
Quantity 5 g
Formula Weight 167.16
Percent Purity ≥97.0% (GC)
Physical Form Crystalline Powder
Chemical Name or Material 2-Methoxy-6-nitrotoluene
EINECSNumber (3)-0799
TSCA No