accessibility menu, dialog, popup

UserName

CH3OH3CNOO

CAS RN 4837-88-1

IUPAC Name: 1-methoxy-2-methyl-3-nitrobenzene
Synonyms: 1-meo-2-ME-3-NO2-benzene
Molecular Weight (g/mol): 167.16
Molecular Formula: C8H9NO3
InChi Key: HQCZLEAGIOIIMC-UHFFFAOYSA-N
SMILES: COC1=CC=CC(=C1C)[N+]([O-])=O
Percent Purity 
  • (2)
Physical Form 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results