missing translation for 'onlineSavingsMsg'
Learn More

2-Methoxy-5-(trifluoromethyl)aniline 98.0+%, TCI America™

Catalog No. M22125G
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
M22125G 5 g
M221225G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. M22125G Supplier TCI America Supplier No. M22125G

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 349-65-5
Molecular Formula C8H8F3NO
Molecular Weight (g/mol) 191.153
MDL Number MFCD00042486
InChI Key RKUSRLUGUVDNKP-UHFFFAOYSA-N
Synonym 2-methoxy-5-trifluoromethyl aniline, 3-amino-4-methoxybenzotrifluoride, 2-amino-4-trifluoromethylanisole, 2-methoxy-5-trifluoromethyl-aniline, 2-methoxy-5-trifluoromethyl-phenylamine, 2-methoxy-5-trifluoromethyl phenylamine, benzenamine, 2-methoxy-5-trifluoromethyl, pubchem2720, acmc-209ic5, 4-trifluromethyl-o-anisidine
PubChem CID 600637
IUPAC Name 2-methoxy-5-(trifluoromethyl)aniline
SMILES COC1=C(C=C(C=C1)C(F)(F)F)N

Specifications

Melting Point 60°C
Quantity 5 g
Formula Weight 191.15
Percent Purity ≥98.0% (GC,T)
Physical Form Crystalline Powder
Chemical Name or Material 2-Methoxy-5-(trifluoromethyl)aniline