accessibility menu, dialog, popup

UserName

CAS RN 349-65-5

H3COH2NFFF

CAS RN 349-65-5

IUPAC Name: 2-methoxy-5-(trifluoromethyl)aniline
Synonyms: 2-methoxy-5-(trifluoromethyl)aniline
Molecular Weight (g/mol): 191.15
Molecular Formula: C8H8F3NO
InChi Key: RKUSRLUGUVDNKP-UHFFFAOYSA-N
SMILES: COC1=CC=C(C=C1N)C(F)(F)F
Percent Purity 
  • (2)
Physical Form 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results