missing translation for 'onlineSavingsMsg'
Learn More

2-Methoxy-5-sulfamoylbenzoic acid, 97%

Catalog No. p-7027158
Change view
Click to view available options
Quantity:
10 g
50 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAB2369809 10 g
AAB2369818 50 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAB2369809 Supplier Thermo Scientific Chemicals Supplier No. B2369809

CAS: 22117-85-7 | C8H9NO5S | 231.222 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 22117-85-7
Molecular Formula C8H9NO5S
Molecular Weight (g/mol) 231.222
MDL Number MFCD00129997
InChI Key SQAILWDRVDGLGY-UHFFFAOYSA-N
Synonym 2-methoxy-5-aminosulfonylbenzoic acid, benzoic acid, 5-aminosulfonyl-2-methoxy, 5-sulphamoyl-o-anisic acid, 2-methoxy-5-sulfonylbenzoic acid, sulpiride impurity d, acmc-209fsc, 2-methoxy-5-sulfamoylbenzoicacid, 2-methoxy-5-sulphamoylbenzoic acid, 2-methoxy-5-sulfamoyl benzoic acid, 2-methoxy-5-sulfamoyl-benzoic acid
PubChem CID 89601
IUPAC Name 2-methoxy-5-sulfamoylbenzoic acid
SMILES COC1=C(C=C(C=C1)S(=O)(=O)N)C(=O)O

Specifications

Quantity 10 g
Formula Weight 232.23
Percent Purity 97%
Chemical Name or Material 2-Methoxy-5-sulfamoylbenzoic acid
Hazard Category H315-H319-H335
Hazard Statement GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
EINECSNumber 244-789-1
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only