accessibility menu, dialog, popup

UserName

CAS RN 22117-85-7

IUPAC Name: 2-methoxy-5-sulfamoylbenzoate
Synonyms: 2-methoxy-5-sulfamoylbenzoate
Molecular Weight (g/mol): 230.21
Molecular Formula: C8H8NO5S
InChi Key: SQAILWDRVDGLGY-UHFFFAOYSA-M
SMILES: COC1=CC=C(C=C1C([O-])=O)S(N)(=O)=O

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results