missing translation for 'onlineSavingsMsg'
Learn More
Learn More
2-Methoxy-5-methylaniline 98.0+%, TCI America™
Supplier: TCI America M0115500G
Specifications
| 2-Methoxy-5-methylaniline | |
| 52°C | |
| C8H11NO | |
| 500 g | |
| WXWCDTXEKCVRRO-UHFFFAOYSA-N | |
| 2-methoxy-5-methylaniline | |
| 8445 | |
| 137.18 | |
| Crystalline Powder |
| 120-71-8 | |
| 235°C | |
| MFCD00007815 | |
| p-cresidine, cresidine, 5-methyl-o-anisidine, para-cresidine, benzenamine, 2-methoxy-5-methyl, krezidin, 2-amino-4-methylanisole, 3-amino-4-methoxytoluene, 4-methoxy-m-toluidine, 6-methoxy-m-toluidine | |
| CC1=CC(=C(C=C1)OC)N | |
| 137.182 | |
| CHEBI:82307 | |
| ≥98.0% (GC,T) |
Chemical Identifiers
| 120-71-8 | |
| 137.182 | |
| WXWCDTXEKCVRRO-UHFFFAOYSA-N | |
| 8445 | |
| 2-methoxy-5-methylaniline |
| C8H11NO | |
| MFCD00007815 | |
| p-cresidine, cresidine, 5-methyl-o-anisidine, para-cresidine, benzenamine, 2-methoxy-5-methyl, krezidin, 2-amino-4-methylanisole, 3-amino-4-methoxytoluene, 4-methoxy-m-toluidine, 6-methoxy-m-toluidine | |
| CHEBI:82307 | |
| CC1=CC(=C(C=C1)OC)N |
Safety and Handling
EINECSNumber : (3)-0614
RTECSNumber : BZ6720000
TSCA : Yes