missing translation for 'onlineSavingsMsg'
Learn More

2-Methoxy-5-methylaniline 98.0+%, TCI America™

Catalog No. p-7131998
Change view
Click to view available options
Quantity:
25 g
500 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
M011525G 25 g
M0115500G 500 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. M011525G Supplier TCI America Supplier No. M011525G
Only null left

Chemical Identifiers

CAS 120-71-8
Molecular Formula C8H11NO
Molecular Weight (g/mol) 137.182
MDL Number MFCD00007815
InChI Key WXWCDTXEKCVRRO-UHFFFAOYSA-N
Synonym p-cresidine, cresidine, 5-methyl-o-anisidine, para-cresidine, benzenamine, 2-methoxy-5-methyl, krezidin, 2-amino-4-methylanisole, 3-amino-4-methoxytoluene, 4-methoxy-m-toluidine, 6-methoxy-m-toluidine
PubChem CID 8445
ChEBI CHEBI:82307
IUPAC Name 2-methoxy-5-methylaniline
SMILES CC1=CC(=C(C=C1)OC)N

Specifications

Melting Point 52°C
Boiling Point 235°C
Quantity 25 g
Formula Weight 137.18
Percent Purity ≥98.0% (GC,T)
Physical Form Crystalline Powder
Chemical Name or Material 2-Methoxy-5-methylaniline
EINECSNumber (3)-0614
RTECSNumber BZ6720000
TSCA Yes