missing translation for 'onlineSavingsMsg'
Learn More

2-Methoxy-4-nitrophenyl isothiocyanate, 97%

Catalog No. AAL1215006
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAL1215006 5 g
AAL1215003 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAL1215006 Supplier Thermo Scientific Chemicals Supplier No. L1215006
Get Up to $1,800 in Chemicals with Purchase
View all promotions

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

CAS: 190774-55-1 | C8H6N2O3S | 210.207 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 190774-55-1
Molecular Formula C8H6N2O3S
Molecular Weight (g/mol) 210.207
MDL Number MFCD00041379
InChI Key NXWXXLFRMVILJN-UHFFFAOYSA-N
Synonym 2-methoxy-4-nitrophenyl isothiocyanate, 1-isothiocyanato-2-methoxy-4-nitro-benzene, acmc-20ans0, 2-methoxy4-nitrophenyl isothiocyanate, 2-methoxy-4-nitrophenylisothiocyanate, #, benzene, 1-isothiocyanato-2-methoxy-4-nitro, benzene, 1-isothiocyanato-2-methoxy-4-nitro-9ci
PubChem CID 610837
IUPAC Name 1-isothiocyanato-2-methoxy-4-nitrobenzene
SMILES COC1=C(C=CC(=C1)[N+](=O)[O-])N=C=S

Specifications

Melting Point 107°C to 109°C
Quantity 5 g
UN Number UN2923
Beilstein 6924925
Sensitivity Moisture sensitive
Formula Weight 210.21
Percent Purity 97%
Chemical Name or Material 2-Methoxy-4-nitrophenyl isothiocyanate
Hazard Category H302+H312+H332-H314-H335
Hazard Statement GHS H Statement
H331-H314-H318-H302-H312
Toxic if inhaled.
Causes severe skin burns and eye damage.
Causes serious eye damage.
Harmful if swallowed.
Harmful in contact with skin.
Precautionary Statement P260-P264b-P270-P271-P280-P303+P361+P353-P304+P340-P305+P351+P338-P310-P330-P331-P363-P501c
DOTInformation Transport Hazard Class: 8; Packing Group: III; Proper Shipping Name: CORROSIVE SOLIDS, TOXIC, N.O.S.
EINECSNumber 000-000-0
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only