missing translation for 'onlineSavingsMsg'
Learn More

2-Methoxy-4-nitrobenzonitrile, 98%

Catalog No. p-7029877
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAH2739106 5 g
AAH2739114 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAH2739106 Supplier Thermo Scientific Chemicals Supplier No. H2739106

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

CAS: 101084-96-2 | C8H6N2O3 | 178.147 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 101084-96-2
Molecular Formula C8H6N2O3
Molecular Weight (g/mol) 178.147
MDL Number MFCD02093781
InChI Key MLIKCKXLGYEGAO-UHFFFAOYSA-N
Synonym 2-cyano-5-nitroanisole, acmc-20a4y1, 4-nitro-2-methoxybenzonitrile, 2-methoxy-4-nitro-benzonitrile, 2-methyloxy-4-nitrobenzonitrile, 2-methoxy-4-nitrobenzonitrile
PubChem CID 291867
IUPAC Name 2-methoxy-4-nitrobenzonitrile
SMILES COC1=C(C=CC(=C1)[N+](=O)[O-])C#N

Specifications

Melting Point 177°C to 180°C
Quantity 5 g
UN Number UN3439
Formula Weight 178.15
Percent Purity 98%
Chemical Name or Material 2-Methoxy-4-nitrobenzonitrile
Hazard Category H302+H312+H332-H315-H319-H335
Hazard Statement GHS H Statement
H331-H302-H312-H315-H319-H335
Toxic if inhaled.
Harmful if swallowed.
Harmful in contact with skin.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement P261-P264b-P270-P271-P280-P301+P312-P302+P352-P304+P340-P305+P351+P338-P312-P330-P332+P313-P362-P501c
DOTInformation Transport Hazard Class: 6.1; Packing Group: III; Proper Shipping Name: NITRILES, SOLID, TOXIC, N.O.S.
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only