accessibility menu, dialog, popup

UserName

CAS RN 101084-96-2

IUPAC Name: 2-methoxy-4-nitrobenzonitrile
Synonyms: 2-methoxy-4-nitrobenzonitrile
Molecular Weight (g/mol): 178.15
Molecular Formula: C8H6N2O3
InChi Key: MLIKCKXLGYEGAO-UHFFFAOYSA-N
SMILES: COC1=C(C=CC(=C1)[N+]([O-])=O)C#N
Molecular Weight (g/mol) 
  • (4)
  • (1)
Percent Purity 
  • (2)
  • (3)
Quantity 
  • (1)
  • (1)
  • (2)
  • (1)
Physical Form 
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (3)
  • (2)
  • (3)
  • (4)
  • (1)
  • (2)
  • (3)
  • (1)
  • (1)
  • (2)
  • (1)
  • (2)