missing translation for 'onlineSavingsMsg'
Learn More
Learn More
2-Methoxy-4-methylphenol 98.0+%, TCI America™
Supplier: TCI America M011425ML
Specifications
| 2-Methoxy-4-methylphenol | |
| 5°C | |
| 221°C | |
| MFCD00002378 | |
| creosol, 4-methylguaiacol, 2-methoxy-p-cresol, homoguaiacol, p-methylguaiacol, phenol, 2-methoxy-4-methyl, p-creosol, 4-methyl guaiacol, 2-methoxy-4-cresol, 4-hydroxy-3-methoxytoluene | |
| CC1=CC(=C(C=C1)O)OC | |
| 138.166 | |
| 138.17 | |
| Liquid |
| 93-51-6 | |
| Yellow | |
| C8H10O2 | |
| 25 mL | |
| PETRWTHZSKVLRE-UHFFFAOYSA-N | |
| 2-methoxy-4-methylphenol | |
| 7144 | |
| ≥98.0% (GC) |
Chemical Identifiers
| 93-51-6 | |
| 138.166 | |
| PETRWTHZSKVLRE-UHFFFAOYSA-N | |
| 7144 | |
| CC1=CC(=C(C=C1)O)OC |
| C8H10O2 | |
| MFCD00002378 | |
| creosol, 4-methylguaiacol, 2-methoxy-p-cresol, homoguaiacol, p-methylguaiacol, phenol, 2-methoxy-4-methyl, p-creosol, 4-methyl guaiacol, 2-methoxy-4-cresol, 4-hydroxy-3-methoxytoluene | |
| 2-methoxy-4-methylphenol |
Safety and Handling
EINECSNumber : (3)-0607&(3)-0635
RTECSNumber : GP1755000
TSCA : Yes