missing translation for 'onlineSavingsMsg'
Learn More

2-Methoxy-4-methylphenol 98.0+%, TCI America™

Catalog No. p-7131997
Change view
Click to view available options
Quantity:
25 mL
100 mL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
M011425ML 25 mL
M0114100ML 100 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. M011425ML Supplier TCI America Supplier No. M011425ML
Only null left

Chemical Identifiers

CAS 93-51-6
Molecular Formula C8H10O2
Molecular Weight (g/mol) 138.166
MDL Number MFCD00002378
InChI Key PETRWTHZSKVLRE-UHFFFAOYSA-N
Synonym creosol, 4-methylguaiacol, 2-methoxy-p-cresol, homoguaiacol, p-methylguaiacol, phenol, 2-methoxy-4-methyl, p-creosol, 4-methyl guaiacol, 2-methoxy-4-cresol, 4-hydroxy-3-methoxytoluene
PubChem CID 7144
IUPAC Name 2-methoxy-4-methylphenol
SMILES CC1=CC(=C(C=C1)O)OC

Specifications

Melting Point 5°C
Color Yellow
Boiling Point 221°C
Quantity 25 mL
Formula Weight 138.17
Percent Purity ≥98.0% (GC)
Physical Form Liquid
Chemical Name or Material 2-Methoxy-4-methylphenol
EINECSNumber (3)-0607&(3)-0635
RTECSNumber GP1755000
TSCA Yes