missing translation for 'onlineSavingsMsg'
Learn More

2-Methoxy-4-methylphenol 98.0+%, TCI America™

Supplier:  TCI America M0114100ML

 View more versions of this product

Catalog No. M0114100ML


Only null left
Explore available promotions

Description

Description

Specifications

Specifications

2-Methoxy-4-methylphenol
5°C
221°C
MFCD00002378
creosol, 4-methylguaiacol, 2-methoxy-p-cresol, homoguaiacol, p-methylguaiacol, phenol, 2-methoxy-4-methyl, p-creosol, 4-methyl guaiacol, 2-methoxy-4-cresol, 4-hydroxy-3-methoxytoluene
CC1=CC(=C(C=C1)O)OC
138.166
138.17
Liquid
93-51-6
Yellow
C8H10O2
100 mL
PETRWTHZSKVLRE-UHFFFAOYSA-N
2-methoxy-4-methylphenol
7144
≥98.0% (GC)

Chemical Identifiers

93-51-6
138.166
PETRWTHZSKVLRE-UHFFFAOYSA-N
7144
CC1=CC(=C(C=C1)O)OC
C8H10O2
MFCD00002378
creosol, 4-methylguaiacol, 2-methoxy-p-cresol, homoguaiacol, p-methylguaiacol, phenol, 2-methoxy-4-methyl, p-creosol, 4-methyl guaiacol, 2-methoxy-4-cresol, 4-hydroxy-3-methoxytoluene
2-methoxy-4-methylphenol
Product Suggestions

Product Suggestions

Videos
Safety and Handling

Safety and Handling

EINECSNumber : (3)-0607&(3)-0635

RTECSNumber : GP1755000

TSCA : Yes

SDS
Documents

Documents

Promotions

Promotions