missing translation for 'onlineSavingsMsg'
Learn More
Learn More
2-Methoxy-4-ethylphenol 98.0+%, TCI America™
Supplier: TCI America E035325ML
Specifications
| 2-Methoxy-4-ethylphenol | |
| Yellow | |
| C9H12O2 | |
| 25 mL | |
| CHWNEIVBYREQRF-UHFFFAOYSA-N | |
| 4-ethyl-2-methoxyphenol | |
| 62465 | |
| ≥98.0% (GC) |
| 2785-89-9 | |
| 235°C | |
| MFCD00038714 | |
| 4-ethylguaiacol, p-ethylguaiacol, 2-methoxy-4-ethylphenol, phenol, 4-ethyl-2-methoxy, homocresol, guaiacyl ethane, 4-ethyl-2-methoxy-phenol, 4-hydroxy-3-methoxy ethylbenzene, unii-c9nfd83bj5, guaiacol, 4-ethyl | |
| CCC1=CC=C(O)C(OC)=C1 | |
| 152.19 | |
| 152.19 | |
| Liquid |
Chemical Identifiers
| 2785-89-9 | |
| 152.19 | |
| CHWNEIVBYREQRF-UHFFFAOYSA-N | |
| 62465 | |
| CCC1=CC=C(O)C(OC)=C1 |
| C9H12O2 | |
| MFCD00038714 | |
| 4-ethylguaiacol, p-ethylguaiacol, 2-methoxy-4-ethylphenol, phenol, 4-ethyl-2-methoxy, homocresol, guaiacyl ethane, 4-ethyl-2-methoxy-phenol, 4-hydroxy-3-methoxy ethylbenzene, unii-c9nfd83bj5, guaiacol, 4-ethyl | |
| 4-ethyl-2-methoxyphenol |
Safety and Handling
EINECSNumber : (3)-0635
TSCA : Yes