missing translation for 'onlineSavingsMsg'
Learn More

2-Methoxy-4-ethylphenol 98.0+%, TCI America™

Catalog No. p-7128955
Change view
Click to view available options
Quantity:
5 mL
25 mL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
E03535ML 5 mL
E035325ML 25 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. E03535ML Supplier TCI America Supplier No. E03535ML

Chemical Identifiers

CAS 2785-89-9
Molecular Formula C9H12O2
Molecular Weight (g/mol) 152.19
MDL Number MFCD00038714
InChI Key CHWNEIVBYREQRF-UHFFFAOYSA-N
Synonym 4-ethylguaiacol, p-ethylguaiacol, 2-methoxy-4-ethylphenol, phenol, 4-ethyl-2-methoxy, homocresol, guaiacyl ethane, 4-ethyl-2-methoxy-phenol, 4-hydroxy-3-methoxy ethylbenzene, unii-c9nfd83bj5, guaiacol, 4-ethyl
PubChem CID 62465
IUPAC Name 4-ethyl-2-methoxyphenol
SMILES CCC1=CC=C(O)C(OC)=C1

Specifications

Color Yellow
Boiling Point 235°C
Quantity 5 mL
Formula Weight 152.19
Percent Purity ≥98.0% (GC)
Physical Form Liquid
Chemical Name or Material 2-Methoxy-4-ethylphenol
EINECSNumber (3)-0635
TSCA Yes