missing translation for 'onlineSavingsMsg'
Learn More
Learn More
2-Methoxy-1-butanol 98.0+%, TCI America™
Supplier: TCI America M149725G
Specifications
| 2-Methoxy-1-butanol | |
| Yellow | |
| C5H12O2 | |
| 1993 | |
| CCC(CO)OC | |
| 104.149 | |
| 104.15 | |
| Liquid |
| 15467-25-1 | |
| 146°C | |
| 25 g | |
| IPUDBCXGMBSQGH-UHFFFAOYSA-N | |
| 2-methoxybutan-1-ol | |
| 85854 | |
| ≥98.0% (GC) |
Chemical Identifiers
| 15467-25-1 | |
| 104.149 | |
| 85854 | |
| CCC(CO)OC |
| C5H12O2 | |
| IPUDBCXGMBSQGH-UHFFFAOYSA-N | |
| 2-methoxybutan-1-ol |
Safety and Handling
EINECSNumber : (7)-0097
TSCA : No