accessibility menu, dialog, popup

UserName

CH3HOOH3C

CAS RN 15467-25-1

IUPAC Name: 2-methoxybutan-1-ol
Synonyms: 2-methoxybutan-1-ol
Molecular Weight (g/mol): 104.15
Molecular Formula: C5H12O2
InChi Key: IPUDBCXGMBSQGH-UHFFFAOYNA-N
SMILES: CCC(CO)OC
Percent Purity 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results