missing translation for 'onlineSavingsMsg'
Learn More

2-Methoxy-1-butanol 98.0+%, TCI America™

Catalog No. p-7132762
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
M14975G 5 g
M149725G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. M14975G Supplier TCI America Supplier No. M14975G

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 15467-25-1
Molecular Formula C5H12O2
Molecular Weight (g/mol) 104.149
InChI Key IPUDBCXGMBSQGH-UHFFFAOYSA-N
PubChem CID 85854
IUPAC Name 2-methoxybutan-1-ol
SMILES CCC(CO)OC

Specifications

Color Yellow
Boiling Point 146°C
Quantity 5 g
UN Number 1993
Formula Weight 104.15
Percent Purity ≥98.0% (GC)
Physical Form Liquid
Chemical Name or Material 2-Methoxy-1-butanol
EINECSNumber (7)-0097
TSCA No