missing translation for 'onlineSavingsMsg'
Learn More
Learn More
2-Mercaptopyridine N-Oxide 95.0+%, TCI America™
$95.06 - $264.70
Chemical Identifiers
| CAS | 1121-31-9 |
|---|---|
| Molecular Formula | C5H5NOS |
| Molecular Weight (g/mol) | 127.161 |
| MDL Number | MFCD00006196 |
| InChI Key | YBBJKCMMCRQZMA-UHFFFAOYSA-N |
| Synonym | pyrithione, 2-mercaptopyridine n-oxide, 2-mercaptopyridine-n-oxide, 1-hydroxy-2-pyridinethione, omadine, 2-pyridinethiol, 1-oxide, 1-hydroxyl-1h-pyridine-2-thione, n-hydroxypyridine-2-thione, 1-hydroxy-2 1h-pyridinethione, 2-pyridinethiol 1-oxide |
| PubChem CID | 1570 |
| ChEBI | CHEBI:36578 |
| IUPAC Name | 1-hydroxypyridine-2-thione |
| SMILES | C1=CC(=S)N(C=C1)O |
Chemical Identifiers
| 1121-31-9 | |
| 127.161 | |
| YBBJKCMMCRQZMA-UHFFFAOYSA-N | |
| 1570 | |
| 1-hydroxypyridine-2-thione |
| C5H5NOS | |
| MFCD00006196 | |
| pyrithione, 2-mercaptopyridine n-oxide, 2-mercaptopyridine-n-oxide, 1-hydroxy-2-pyridinethione, omadine, 2-pyridinethiol, 1-oxide, 1-hydroxyl-1h-pyridine-2-thione, n-hydroxypyridine-2-thione, 1-hydroxy-2 1h-pyridinethione, 2-pyridinethiol 1-oxide | |
| CHEBI:36578 | |
| C1=CC(=S)N(C=C1)O |
Specifications
| 1121-31-9 | |
| White-Yellow | |
| MFCD00006196 | |
| pyrithione, 2-mercaptopyridine n-oxide, 2-mercaptopyridine-n-oxide, 1-hydroxy-2-pyridinethione, omadine, 2-pyridinethiol, 1-oxide, 1-hydroxyl-1h-pyridine-2-thione, n-hydroxypyridine-2-thione, 1-hydroxy-2 1h-pyridinethione, 2-pyridinethiol 1-oxide | |
| C1=CC(=S)N(C=C1)O | |
| 127.161 | |
| CHEBI:36578 | |
| ≥95.0% (T) | |
| 2-Mercaptopyridine N-Oxide |
| 72°C | |
| C5H5NOS | |
| 5 g | |
| YBBJKCMMCRQZMA-UHFFFAOYSA-N | |
| 1-hydroxypyridine-2-thione | |
| 1570 | |
| 127.16 | |
| Crystalline Powder |
Safety and Handling
EINECSNumber : (5)-0752
RTECSNumber : UT8999000
TSCA : Yes
Recommended Storage : Refrigerator