missing translation for 'onlineSavingsMsg'
Learn More

2-Mercaptopyridine N-Oxide 95.0+%, TCI America™

Supplier:  TCI America H03025G

 View more versions of this product

Catalog No. H03025G


Only null left
Explore available promotions

Description

Description

Specifications

Specifications

2-Mercaptopyridine N-Oxide
72°C
C5H5NOS
5 g
YBBJKCMMCRQZMA-UHFFFAOYSA-N
1-hydroxypyridine-2-thione
1570
127.16
Crystalline Powder
1121-31-9
White-Yellow
MFCD00006196
pyrithione, 2-mercaptopyridine n-oxide, 2-mercaptopyridine-n-oxide, 1-hydroxy-2-pyridinethione, omadine, 2-pyridinethiol, 1-oxide, 1-hydroxyl-1h-pyridine-2-thione, n-hydroxypyridine-2-thione, 1-hydroxy-2 1h-pyridinethione, 2-pyridinethiol 1-oxide
C1=CC(=S)N(C=C1)O
127.161
CHEBI:36578
≥95.0% (T)

Chemical Identifiers

1121-31-9
127.161
YBBJKCMMCRQZMA-UHFFFAOYSA-N
1570
1-hydroxypyridine-2-thione
C5H5NOS
MFCD00006196
pyrithione, 2-mercaptopyridine n-oxide, 2-mercaptopyridine-n-oxide, 1-hydroxy-2-pyridinethione, omadine, 2-pyridinethiol, 1-oxide, 1-hydroxyl-1h-pyridine-2-thione, n-hydroxypyridine-2-thione, 1-hydroxy-2 1h-pyridinethione, 2-pyridinethiol 1-oxide
CHEBI:36578
C1=CC(=S)N(C=C1)O
Product Suggestions

Product Suggestions

Videos
Safety and Handling

Safety and Handling

EINECSNumber : (5)-0752

RTECSNumber : UT8999000

TSCA : Yes

Recommended Storage : Refrigerator

SDS
Documents

Documents

Promotions

Promotions