accessibility menu, dialog, popup

UserName

OHNS

CAS RN 1121-31-9

IUPAC Name: 1-hydroxy-1,2-dihydropyridine-2-thione
Synonyms: pyrithione pyrithione sodium salt 2-hpt cpd pyrithione sodium pyrithione barium (2:1) salt omadine 67Ga-MPO pyrithione potassium salt 68ga-mercaptopyridine-n-oxide pyrithione magnesium salt
Molecular Weight (g/mol): 127.16
Molecular Formula: C5H5NOS
InChi Key: YBBJKCMMCRQZMA-UHFFFAOYSA-N
SMILES: ON1C=CC=CC1=S
Molecular Weight (g/mol) 
  • (3)
  • (5)
Percent Purity 
  • (2)
  • (6)
Quantity 
  • (1)
  • (1)
  • (2)
  • (1)
  • (3)
Physical Form 
  • (2)
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (6)
  • (3)
  • (5)
  • (2)
  • (6)
  • (1)
  • (1)
  • (2)
  • (1)
  • (3)
  • (2)
  • (2)