missing translation for 'onlineSavingsMsg'
Learn More

2-Mercaptobenzyl alcohol, tech. 90%

CAS: 4521-31-7 | C7H8OS | 140.2 g/mol

Supplier:  Thermo Scientific Chemicals L1153606

 View more versions of this product

Catalog No. AAL1153606

Only null left
Explore available promotions

Description

Description

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Specifications

2-Mercaptobenzyl alcohol
31°C to 32°C
94°C to 96°C (0.2 mmHg)
Like Rotten Eggs
1.623
5 g
2-mercaptobenzyl alcohol, 2-mercaptophenyl methanol, o-mercaptobenzyl alcohol, 2-sulfanylphenyl methanol, 2-mercaptobenzylalcohol, 2-hydroxymethyl thiophenol, benzenemethanol, 2-mercapto, hydroxymethylthiophenol, o-mercaptobenzylalcohol, acmc-1csoz
C1=CC=C(C(=C1)CO)S
140.2
140.21
Technical
4521-31-7
1.21
>110°C (230°F)
C7H8OS
MFCD00014448
2206253
FYWFCRHZXORPFH-UHFFFAOYSA-N
(2-sulfanylphenyl)methanol
138278
90%

Chemical Identifiers

4521-31-7
140.2
FYWFCRHZXORPFH-UHFFFAOYSA-N
138278
C1=CC=C(C(=C1)CO)S
C7H8OS
MFCD00014448
2-mercaptobenzyl alcohol, 2-mercaptophenyl methanol, o-mercaptobenzyl alcohol, 2-sulfanylphenyl methanol, 2-mercaptobenzylalcohol, 2-hydroxymethyl thiophenol, benzenemethanol, 2-mercapto, hydroxymethylthiophenol, o-mercaptobenzylalcohol, acmc-1csoz
(2-sulfanylphenyl)methanol
Product Suggestions

Product Suggestions

Videos
Safety and Handling

Safety and Handling

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

P280-P305+P351+P338-P310

H318

DOTInformation : Hazard Class: 9; Packaging Group: III

EINECSNumber : 000-000-0

TSCA : No

Recommended Storage : Ambient temperatures

SDS
Documents

Documents

Promotions

Promotions

RUO – Research Use Only