missing translation for 'onlineSavingsMsg'
Learn More

2-Mercaptobenzyl alcohol, tech. 90%

Catalog No. AAL1153614
Change view
Click to view available options
Quantity:
1 g
5 g
25 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAL1153614 25 g
AAL1153603 1 g
AAL1153606 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. AAL1153614 Supplier Thermo Scientific Chemicals Supplier No. L1153614
Get Up to $1,800 in Chemicals with Purchase
View all promotions

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

CAS: 4521-31-7 | C7H8OS | 140.2 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 4521-31-7
Molecular Formula C7H8OS
Molecular Weight (g/mol) 140.2
MDL Number MFCD00014448
InChI Key FYWFCRHZXORPFH-UHFFFAOYSA-N
Synonym 2-mercaptobenzyl alcohol, 2-mercaptophenyl methanol, o-mercaptobenzyl alcohol, 2-sulfanylphenyl methanol, 2-mercaptobenzylalcohol, 2-hydroxymethyl thiophenol, benzenemethanol, 2-mercapto, hydroxymethylthiophenol, o-mercaptobenzylalcohol, acmc-1csoz
PubChem CID 138278
IUPAC Name (2-sulfanylphenyl)methanol
SMILES C1=CC=C(C(=C1)CO)S

Specifications

Melting Point 31°C to 32°C
Density 1.21
Boiling Point 94°C to 96°C (0.2 mmHg)
Flash Point >110°C (230°F)
Odor Like Rotten Eggs
Refractive Index 1.623
Quantity 25 g
Beilstein 2206253
Formula Weight 140.21
Percent Purity 90%
Grade Technical
Chemical Name or Material 2-Mercaptobenzyl alcohol
Show More Show Less
Hazard Category H318
Hazard Statement GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement P280-P305+P351+P338-P310
DOTInformation Hazard Class: 9; Packaging Group: III
EINECSNumber 000-000-0
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only