missing translation for 'onlineSavingsMsg'
Learn More

2-(Isopropylsulfonyl)aniline 98.0+%, TCI America™

Catalog No. p-7131722
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
I10331G 1 g
I10335G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. I10331G Supplier TCI America Supplier No. I10331G

Chemical Identifiers

CAS 76697-50-2
Molecular Formula C9H13NO2S
Molecular Weight (g/mol) 199.27
MDL Number MFCD08445603
InChI Key GMLAMRMKROYXNZ-UHFFFAOYSA-N
Synonym 2-Aminophenyl Isopropyl Sulfone
PubChem CID 12646366
IUPAC Name 2-(propane-2-sulfonyl)aniline
SMILES CC(C)S(=O)(=O)C1=CC=CC=C1N

Specifications

Melting Point 85°C
Quantity 1 g
Formula Weight 199.27
Percent Purity ≥98.0% (GC)
Physical Form Crystalline Powder
Chemical Name or Material 2-(Isopropylsulfonyl)aniline