accessibility menu, dialog, popup

UserName

H3CCH3SOOH2N

CAS RN 76697-50-2

IUPAC Name: 2-(propane-2-sulfonyl)aniline
Synonyms: 2-(propane-2-sulfonyl)aniline
Molecular Weight (g/mol): 199.27
Molecular Formula: C9H13NO2S
InChi Key: GMLAMRMKROYXNZ-UHFFFAOYSA-N
SMILES: CC(C)S(=O)(=O)C1=CC=CC=C1N
Percent Purity 
  • (2)
Physical Form 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results