missing translation for 'onlineSavingsMsg'
Learn More

(2-Isopropyl-5-methylphenoxy)acetic Acid 98.0+%, TCI America™

Catalog No. p-7131716
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
I10141G 1 g
I10145G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. I10141G Supplier TCI America Supplier No. I10141G

Chemical Identifiers

CAS 5333-40-4
Molecular Formula C12H16O3
Molecular Weight (g/mol) 208.257
MDL Number MFCD00021757
InChI Key MURZAHIYMVFXCF-UHFFFAOYSA-N
PubChem CID 220116
IUPAC Name 2-(5-methyl-2-propan-2-ylphenoxy)acetic acid
SMILES CC1=CC(=C(C=C1)C(C)C)OCC(=O)O

Specifications

Melting Point 151°C
Color White-Yellow
Quantity 1 g
Formula Weight 208.26
Percent Purity ≥98.0% (GC,T)
Physical Form Crystalline Powder
Chemical Name or Material (2-Isopropyl-5-methylphenoxy)acetic Acid